Learn More
Medchemexpress LLC Quercetin 3-O-β-D-glucofuranoside | 21637-25-2 | 464.38 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN076810MM/1ML
Encompass_Preferred
Quercetin 3-O-β-D-glucofuranoside is a compound found widely in plants and is classified as a flavonoid and flavonol. It acts as an effective antioxidant and has been shown to suppress eosinophilic inflammation. Studies suggest it can blunt the negative effects of hydrogen peroxide on certain cells and reduce eosinophil counts in experimental settings.
  • It is an effective antioxidant.
  • It functions as an eosinophilic inflammation suppressor.
  • It has been observed to counteract oxidative-induced damage in RGC-5 cells.
  • It lowers eosinophil counts in bronchoalveolar lavage fluid, blood, and lung parenchyma in animal models.
  • It reduces neutrophil counts in blood and interleukin-5 levels in lung homogenate in experimental models.
Catalog No. 50-004-17128