Learn More
Cayman Chemical QuercetIn 3ODGlucose7O 10mg
SDP
Supplier:  Cayman Chemical 3613010 MG
Encompass_Preferred
A flavonoid glycoside with lipid-lowering activity; reduces sodium oleate-induced increases in triglyceride levels in HepG2 cells at 30 µM
Catalog No. 50-291-1820