Learn More
Cayman Chemical QuercetIn-7-OD-Glucopyrno 10mg
SDP
Supplier:  Cayman Chemical 2764110 MG
Encompass_Preferred
A flavonoid; has antioxidant activity in a ORAC assay and decreases tert-butyl hydroperoxide-induced ROS in L-929 cells at 0.25 and 1 UG/ml; reduces protein levels of iNOS and COX-2 in LPS-stimulated RAW 264.7 cells at 15 and 30 UG/ml; decreases angiogenesis in isolated rat aortic rings and proliferation of HUVECs but has no effect on tube formation or chemotaxis of HUVECs at 100 µM
Catalog No. 50-294-1500