Learn More
Cayman Chemical Quercetin hydrate
SDP
Supplier:  Cayman Chemical 1000516950 G
Encompass_Preferred
A flavonoid; reduces autophagy, decreases the levels of ROS and MDA content, and increases total antioxidant capacity in the kidney in a mouse model of cadmium-induced autophagy at 5-100 mg/kg; reduces tumor growth in an HL60 mouse xenograft model at 120 mg/kg every four days; inhibits histamine release from antigen-stimulated RBL-2H3 cells and decreases the expression of pro-inflammatory cytokines in HMC-1 cells
Catalog No. 50-289-0758