Learn More
Medchemexpress LLC Quercetin (standard) | 117-39-5 | 99.6% | 302.24 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY18085R
Encompass_Preferred
Quercetin (Standard) is the analytical standard of Quercetin, a natural flavonoid that acts as a stimulator of recombinant SIRT1 and a PI3K inhibitor. This compound is used as a reference standard for assays and is commonly employed in qualitative, quantitative, and methodological research experiments.
  • Intended for research and analytical applications
  • Acts as a stimulator of recombinant SIRT1
  • Functions as a PI3K inhibitor
  • Suitable for HPLC, GC, and MS experiments
Catalog No. 50-003-84182