Learn More
Aat Bioquest Fluo-8, AM | 1mg
SDP
Supplier:  Aat Bioquest 21080
Encompass_Preferred
Calcium measurements are critical for numerous biological investigations. Fluorescent probes that show spectral responses upon binding Ca2+ have enabled researchers to investigate changes in intracellular free Ca2+ concentrations by using fluorescence microscopy, flow cytometry, fluorescence spectroscopy, and fluorescence microplate readers. Fluo-3 AM and Fluo-4 AM are most commonly used among the visible light-excitable calcium indicators for live-cell calcium imaging. However, Fluo-3 AM and Fluo-4 AM are only moderately fluorescent in live cells upon esterase hydrolysis and require harsh cell loading conditions to maximize their cellular calcium responses. Fluo-8 dyes are developed to improve cell loading and calcium response while maintaining the convenient Fluo-3 and Fluo-4 spectral wavelengths of Ex/Em = ∼490/∼520 nm. Fluo-8 AM can be loaded into cells at room temperature, while Fluo-3 AM and Fluo-4 AM require 37°C for cell loading.
Catalog No. NC0027049