Learn More
Medchemexpress LLC HY-B0031 500mg Medchemexpress, Quetiapine (hemifumarate) CAS:111974-72-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0031500MG
Encompass_Preferred
Medchemexpress, HY-B0031 500mg Quetiapine (hemifumarate) CAS:111974-72-2 Quetiapine hemifumarate is a 5-HT receptors agonist with a pEC50 of 4.77 for human 5-HT1A receptor. Quetiapine hemifumarate is a dopamine receptor antagonist with a pIC50 of 6.33 for human D2 receptor. Quetiapine hemifumarate has moderate to high affinity for the human D2, HT1A, 5-HT2A, 5-HT2C receptor with pKis of 7.25, 5.74, 7.54, 5.55. Antidepressant and anxiolytic effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2350