Learn More
Medchemexpress LLC Quetiapine impurity 1 | 1076199-40-0 | MFCD11977896 | 399.51 | C21H25N3O3S | 5 MG

Supplier: Medchemexpress LLC HY1000675MG
Quetiapine impurity 1 is a synthetic impurity related to the antipsychotic quetiapine, supplied for analytical reference and synthetic intermediate applications. Identifiers provided by the manufacturer include CAS 1076199-40-0, molecular formula C21H25N3O3S, and molecular weight 399.51 g·mol⁻¹.
- Used as an analytical reference and drug intermediate
- Supplied as a powder for laboratory handling and storage
- Molecular formula C21H25N3O3S
- Molecular weight 399.51 g·mol⁻¹
- CAS number 1076199-40-0 for unambiguous identification
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.