Learn More
Medchemexpress LLC Quetiapine N-oxide | 1076199-40-0 | MFCD11977896 | >95.0% | 399.51 | C21H25N3O3S | 100 MG

Supplier: Medchemexpress LLC HY100067100MG
Quetiapine impurity 1 (Quetiapine N-oxide) is an analytical impurity reference used for identification and quantification of degradation products, metabolites, and related impurities in quetiapine drug substances and formulations. It is provided as a characterized small-molecule standard for laboratory analytical workflows and method development.
- Analytical reference impurity for quetiapine
- CAS number 1076199-40-0
- Chemical formula C21H25N3O3S
- Molecular weight 399.51 g/mol
- Purity greater than 95% (HPLC)
- Intended for impurity identification and method development
- Supplied in measured quantity suitable for analytical use
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.