Learn More
Medchemexpress LLC Quetiapine N-oxide | 1076199-40-0 | MFCD11977896 | ≥98.0% | 399.51 g/mol | C21H25N3O3S | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10006725MG
Encompass_Preferred
Quetiapine impurity 1 (Quetiapine N-oxide) is a characterized impurity reference standard used in analytical and research applications to identify and quantify quetiapine-related impurities. It is provided as a solid reference material to support method development, impurity profiling, and assay validation. For research use only.
  • Characterized impurity for quetiapine analytical testing.
  • High purity (≥98%) suitable for accurate quantification.
  • Known chemical identity and CAS for unambiguous identification.
  • Compatible with chromatographic and spectrometric methods.
  • Supplied as a stable solid reference material for laboratory use.
Catalog No. 50-002-15203