Learn More
Chemglass Life Sciences Quick Disconnect BRASS Coupling Set: Coupler and Nipple
SDP
Supplier:  Chemglass Life Sciences CG1969M70
Encompass_Preferred
Quick Disconnect BRASS Coupling Set: Coupler and Nipple, with each side having a M16 x 1 Male Thread fitting. Temp Range -50 to 140°C
Catalog No. 50-872-390