Learn More
Sigma Aldrich Fine Chemicals Biosciences Quick Press KBr pellet kit
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z5067611KT
Encompass
This hand press is for making transparent KBr pellets (discs). The pellet is formed using only the pressure of a firm handshake applied steadily to the handle. Kit includes a die set for making 7mm diam. pellets.. Die sets made of hardened nickel based alloy. Anvil faces are optically polished. Optional 2 and 4mm die sets and collarsKBr Pellet Technique1. KBr powder/sample matrix is poured into the hole in the SS collar into which the shorter of the two anvils has first been inserted.2. The longer second anvil is then inserted in the collar and the assembly is placed in the press handle.3. Pressure is applied to the sample by squeezing the handle.4. When the anvils are removed from the collar the KBr pellet is retained inside of the collar.5. The collar is placed in the optional V-shaped sample holder that will fit standard 2 x 3in. slide mount of any spectrometer.6. After analysis the pellet is rinsed out of the collar with water or ejected and saved.
Catalog No. 50-290-8536