Learn More
Zymo Research Corporation Quick-RNA Whole-Blood (50 preps)
SDP
Supplier:  Zymo Research Corporation R1201
Encompass_Preferred
The Quick-RNA Whole Blood kit utilizes DNA/RNA Shield™, a unique preservation and lysis technology, to enable rapid isolation of total RNA from whole or partitioned blood or a cell pellet (after red blood cell lysis). The procedure uses Zymo-Spin column technology in which the sample is pre-filtered on the Zymo-Spin IIICG Column and then purified and concentrated on the Zymo-Spin IC Column. RNA is eluted into 6 l of RNase-free water and is ready for any downstream application including RT-PCR, sequencing, etc.
Catalog No. 50-125-1711