Learn More
Agilent Technologies QUICKCHNGE MULTISITE KIT 10RXN
Supplier:  Agilent Technologies 200515
Encompass
Kit; Mutagenesis; QuikChange Multi Site Directed; 10 rxn (Academic), Contains: DNA polymerase blend, template digestion enzyme, competent cells, positive controls, 10 rxn
Catalog No. NC9331380