Learn More
Sartorius Quickseal Protective Silicone Cap 1/2" (12.7mm)
Supplier:  Sartorius QS08CAPSILNT/DEL
Encompass
This item has a minimum order quantity of 50 per supplier requirements.
The Quickseal protective caps can be installed after collars are cut. The caps are made from flexible and durable silicone. They are suitable for a wide range of temperatures.
Catalog No. NC2194924