Learn More
ILEX LIFE SCIENCES LLC QUICKZYME HUM MMP14 4X96 ASSAY
SDP
Supplier:  ILEX LIFE SCIENCES LLC QZBMMP14HX4
Encompass_Preferred
QuickZyme Human MMP-14 Activity Assay Kit, 4 x 96 Assays, 96-well plate format, immunoassay for active matrix metalloproteinase 14 (cell extracts, tissue homogenates, tissue extracts samples). The QuickZyme Human MMP-14 Activity Assay is an enzyme activity immunoassay that enables quantitative measurement of active human matrix metalloproteinase-14 in cell extracts, tissue homogenates, or tissue extract samples. This 96-well plate format assay is based on the QuickZyme technology, using a modified pro-enzyme as a substrate, which upon activation is able to release color from a chromogenic peptide substrate. This amplification step provides a unique assay sensitivity. ASSAY RANGE: 0.1 - 16 ng/ml. SENSITIVITY: 0.5 ng/ml (2 h incubation); 0.1 ng/ml (5 h incubation). SYNONYMS: MMP14, MMP-X1, Membrane-type matrix metalloproteinase 1, MT-MMP 1, MTMMP1, Membrane-type-1 matrix metalloproteinase, MT1-MMP, MT1MMP.
Catalog No. 50-273-3626