Learn More
ILEX LIFE SCIENCES LLC QUICKZYME MOUSE MMP9 4X96ASSAY
SDP
Supplier:  ILEX LIFE SCIENCES LLC QZBMMP9MX4
Encompass_Preferred
QuickZyme Mouse MMP-9 Activity Assay Kit, 4 x 96 Assays, 96-well plate format. The QuickZyme Mouse MMP-9 Activity Assay is an enzyme activity immunoassay that enables you to specifically measure both endogenous active mouse MMP-9 and total active MMP-9 (following activation of pro-MMP-9 on the plate with APMA). It can be used for the measurement of MMP-8 activity in various biological samples, such as cell culture conditioned medium, serum, plasma, urine and tissue homogenates. This 96-well plate format assay is based on the QuickZyme technology, using a modified pro-enzyme as a substrate, which upon activation is able to release color from a chromogenic peptide substrate. This multiplication step provides a unique assay sensitivity. ASSAY RANGE: 4 - 4000 pg/ml. SENSITIVITY: 20 pg/ml (1 h incubation); 1 pg/ml (6 h incubation). SYNONYMS: Murine matrix metalloproteinase-9, MMP9, 92 kDa gelatinase, 92 kDa type IV collagenase, Gelatinase B, GELB.
Catalog No. 50-273-3621