Learn More
Hampton Research QUIK SCREEN NOA2 185ML
SDP
Supplier:  Hampton Research HR2921A2
Encompass
HR2-921-A2/Quik Screen #A2/Grid Screen Salt HT (E02), 185 ml - 0.8 M Sodium phosphate monobasic monohydrate / Potassium phosphate dibasic pH 5.6. Please note items from this supplier are non-returnable.
Catalog No. 50-180-1