Learn More
Hampton Research QUIK SCREEN NOA4 185ML
SDP
Supplier:  Hampton Research HR2921A4
Encompass
HR2-921-A4/Quik Screen #A4/Grid Screen Salt HT (E04), 185 ml - 0.8 M Sodium phosphate monobasic monohydrate / Potassium phosphate dibasic pH 6.9. Please note items from this supplier are non-returnable.
Catalog No. 50-180-3