Learn More
Agilent Technologies QUIKCHANGE XL MUTAGENESIS KIT
Supplier:  Agilent Technologies 200516
Encompass
QuikChange XL Site-Directed Mutagenesis Kit 1 kit 30 Reactions The kit provides rapid and reliable method for site directed mutagenesis for large constructs For use with large (>8 kb) or difficult constructs
Catalog No. 50-125-161