Learn More
Cayman Chemical QuIlaIc AcId 5mg
SDP
Supplier:  Cayman Chemical 315125 MG
Encompass_Preferred
A triterpene saponin with diverse biological activities; cytotoxic to SNU-1 and KATO III gastric cancer cells (IC50s = 13.6 and 67 µM, respectively); induces hot plate analgesia in mice (ED50 = 20.7 mg/kg); reduces arachidonic acid- or TPA-induced EAr edema in mice when administered at 0.7 and 1.6 mg/ear, respectively
Catalog No. 50-299-0968