Learn More
Medchemexpress LLC Quillaic acid | 631-01-6 | 486.68 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN083950MG
Encompass_Preferred
Quillaic acid is a compound with anti-gastric cancer and anti-proliferation properties, known to promote apoptosis in cancer cells. It also exhibits analgesic and local anti-inflammatory activity, making it relevant for various biological studies.
  • Promotes apoptosis of cancer cells.
  • Exhibits analgesic and local anti-inflammatory activity.
  • Shows inhibitory effect on SNU1 and KATO III cells.
  • Induces dose-dependent analgesia in mouse models.
  • White to off-white solid appearance.
  • Purity of 99.10%.
Catalog No. 50-004-16282