Learn More
Medchemexpress LLC Quillaic acid (Quillaja sapogenin) | 631-01-6 | 486.68 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYN0839100MG
Encompass_Preferred
Quillaic acid (Quillaja sapogenin) is an anti-gastric cancer and anti-proliferation agent that can promote apoptosis of cancer cells. Quillaic acid also has analgesic and local anti-inflammatory activity.
  • Shows inhibitory effect on SNU1 cells and KATO III cells.
  • Induced dose-dependent analgesia in mouse models.
  • Exhibits dose-dependent antinociceptive effects in local tail-flick and whole-body hot plate tests.
Catalog No. 50-004-16377