Learn More
Selleck Chemical LLC Quinacrine 2HCl S4255-1g
SDP
Supplier:  Selleck Chemical LLC S42551G
Encompass_Preferred
Quinacrine 2HCl(Quinacrine dihydrochloride) is a lipophilic cationic drug with multiple actions that is commonly used as an anti-protozoal agent Quinacrine is an effective phospholipase A2 inhibitor
Catalog No. 50-310-9021