Learn More
Medchemexpress LLC Quinagolide hydrochloride | 94424-50-7 | 99.6% | 432.02 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13736A50MG
Encompass_Preferred
Quinagolide hydrochloride is a selective and orally active dopamine D2 receptor agonist. It acts as an inhibitor of prolactin and down-regulates AKT levels and its phosphorylation. It shows antitumor effects and can be used for cancer research.
  • Selective and orally active dopamine D2 receptor agonist.
  • Inhibitor of prolactin.
  • Down-regulates AKT levels and its phosphorylation.
  • Exhibits antitumor effects.
  • Used for cancer research.
  • Reduces non-ergot dopamine receptor 2 (DRD2) mRNA expression.
  • Inhibits invasive properties of endometrial mesenchymal stromal cells.
  • Reduces endothelial differentiation of endometrial mesenchymal stromal cells.
  • Effectively inhibits tumor growth in vivo.
  • Induces normalization of plasma PRL levels and reduced tumor size.
Catalog No. 50-003-52151