Learn More
Apexbio Technology LLC Quinapril HCl 82586-55-8 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B22075.1
Encompass_Preferred
Quinapril hydrochloride (CAS 82586-55-8) is an esterified precursor that undergoes hepatic conversion to quinaprilat an active inhibitor of angiotensin-converting enzyme (ACE) By impeding the conversion of angiotensin I to angiotensin II quinapril modulates the renin-angiotensin-aldosterone system resulting in decreased vasoconstriction and reduced aldosterone secretion This molecule is frequently utilized in cardiovascular research to investigate mechanisms underlying hypertension heart failure and the biological effects of ACE inhibition on vascular and renal systems
Catalog No. 50-000-88129