Learn More
Cayman Chemical QuInaprIlat 10mg
SDP
Supplier:  Cayman Chemical 3600710 MG
Encompass_Preferred
An ACE inhibitor (IC50 = 0.06 nM) and active metabolite of quinapril; inhibits LPC-induced calcium overload and rod to round cell shape change in isolated rat cardiomyocytes at 20 and 50 µM; reduces the frequency of ventricular fibrillation in a dog model of myocardial ischemia and reperfusion injury at 3 UG/kg; reduces blood pressure and proteinuria in rat models of uranyl nitrate- or 5/6 nephrectomy-induced chronic renal failure at 0.1 mg/kg
Catalog No. 50-291-0622