Learn More
Medchemexpress LLC Quinidine (15% dihydroquinidine) | 56-54-2 | 98.4% | 100 G

Supplier: Medchemexpress LLC HYB1751100G
Quinidine (15% dihydroquinidine) is an antiarrhythmic agent. It is a potent, orally active, selective cytochrome P450db inhibitor. Quinidine also acts as a K+ channel blocker with an IC50 of 19.9 μM and can induce apoptosis. It can be used for malaria research.
- Molecular weight: 324.42
- Formula: C20H24N2O2
- Appearance: Solid
- Color: White to off-white
- Structure classification: Alkaloids, Quinoline Alkaloids
- Initial source: Plants other families
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.