Learn More
Medchemexpress LLC Quinidine (15% dihydroquinidine) | 56-54-2 | 98.4% | 50 G
SDP
Supplier:  Medchemexpress LLC HYB175150G
Encompass_Preferred
Quinidine (15% dihydroquinidine) is an antiarrhythmic agent and a potent, orally active, selective cytochrome P450db inhibitor. It also acts as a K+ channel blocker with an IC50 of 19.9 μM and can induce apoptosis. This product is suitable for malaria research and has shown cytotoxicity against MES-SA cells, as well as effects on the PTZ-induced seizure threshold.
  • Acts as an antiarrhythmic agent
  • Potent, orally active, selective cytochrome P450db inhibitor
  • Functions as a K+ channel blocker
  • Capable of inducing apoptosis
  • Applicable for malaria research
  • Exhibits cytotoxicity against MES-SA cells
  • Affects PTZ-induced seizure threshold
Catalog No. 50-004-12802