Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Quinidine methiodide | 42982-87-6 | C21H27IN2O2 | 25 MG
SDP

Supplier:  Medchemexpress LLC 25MGHY150123

Encompass_Preferred

Quinidine methiodide is a metabolite of Quinidine, an antiarrhythmic agent, intended for research use only. This high-purity product, appearing as a white to light yellow solid, requires specific storage conditions to maintain its integrity.

  • Metabolite of Quinidine, an antiarrhythmic agent
  • For research use only
  • Purity of 99.75%
  • Solid, white to light yellow appearance
  • Optimal storage at -20°C under nitrogen, away from moisture
  • Solvent storage at -80°C for 6 months or -20°C for 1 month under nitrogen, away from moisture

Catalog No. 50-003-74539


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.