Learn More
Medchemexpress LLC Quinidine methiodide | 42982-87-6 | C21H27IN2O2 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY15012350MG
Encompass_Preferred
Quinidine methiodide is a metabolite of Quinidine, an antiarrhythmic agent. This product is intended for research use only.
  • High purity: 99.75%
  • Available in solid form
  • Soluble in DMSO for in vitro studies
  • Soluble in various mixtures for in vivo studies
  • Store solid at -20°C under nitrogen
Catalog No. 50-003-75705