Learn More
Selleck Chemical LLC Quinine HCl Dihydrate 10mg 6119-47-7
SDP
Supplier:  Selleck Chemical LLC S250210MG
Encompass_Preferred
Quinine HCl Dihydrate is a white crystalline K+ channel blocker, used to treat malaria. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-526-5