Learn More
Abcam Quinine hemisulfate monohydrate, Potassium channel blocker, 25G
Supplier:  Abcam AB14632725G
Encompass_Preferred
MW 391.47 g/mol. Potassium channel blocker. Antimalarial, anticholinergic, antihypertensive and hypoglycemic agent. Inhibits mitochondrial ATP-regulated potassium channel. Also used to study the metabolism of biocrystalized heme, hemozoin, in malarial parasites and to study the toxicity of heme (FP)-complexes.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4715