Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Quininic acid | 86-68-0 | 250 MG
SDP

Supplier:  Medchemexpress LLC 250MGHYN7354

Encompass_Preferred

Quininic acid, purified from Eucalyptus globulus, cinchona bark, and other plant products, is the most abundant organic acid. It represents up to 86.3% of all organic acids and acts as an astringent and starting material for the synthesis of new pharmaceuticals. It is intended for research use only.

  • Purified from Eucalyptus globulus, cinchona bark, and other plant products
  • Most abundant organic acid, representing up to 86.3% of all organic acids
  • Acts as an astringent
  • Starting material for the synthesis of new pharmaceuticals
  • Solid form, white to yellow color
  • High purity of 99.94%

Catalog No. 50-004-21565


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.