Learn More
Medchemexpress LLC Quinoline-2-carboxaldehyde | 5470-96-2 | 157.17 | 10 G

Supplier: Medchemexpress LLC HYW00816810G
Quinoline-2-carboxaldehyde is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is for research use only and not sold to patients.
- Purity of 99.99%
- Molecular formula C10H7NO
- Molecular weight 157.17
- Storage at -20°C, sealed, away from moisture
- Appearance as a light yellow to yellow solid
- Melting point 66-71°C
- Boiling point 314.3°C at 760 mmHg
- Flash point 151.9°C
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.