Learn More
Medchemexpress LLC Quinoline-2-carboxylic acid | 93-10-7 | 173.17 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW002011
Encompass_Preferred
Quinoline-2-carboxylic acid exhibits antidiabetic activity and serves as a versatile drug intermediate for synthesizing various active compounds.
  • Targets glycosidase and amylases.
  • Relevant for metabolic disease research.
  • Classified under ketones, aldehydes, and acids.
  • Functions as an endogenous metabolite.
  • Appears as a white to off-white solid.
Catalog No. 50-004-48262