Learn More
Medchemexpress LLC Quinoline-2-carboxylic acid | 93-10-7 | 173.17 | 50 G
SDP
Supplier:  Medchemexpress LLC HYW00201150G
Encompass_Preferred
Quinoline-2-carboxylic acid exhibits antidiabetic activity and can be used as a drug intermediate for the synthesis of various active compounds. It reduces the intestinal absorption of carbohydrates by inhibiting α-glucosidase and α-amylase (IC50 are 9.1 μg/mL and 15.5 μg/mL, respectively), thereby lowering postprandial blood glucose levels. This compound can also be used as a drug intermediate for the synthesis of compounds with antiallergic, apoptosis-inducing, or antiproliferative activities.
  • Exhibits antidiabetic activity by reducing intestinal carbohydrate absorption.
  • Inhibits α-glucosidase and α-amylase.
  • Can be used as a drug intermediate for synthesis of compounds with antiallergic, apoptosis-inducing, or antiproliferative activities.
  • Acts as a human endogenous metabolite.
Catalog No. 50-004-45918