Learn More
Medchemexpress LLC Quinoline-2-carboxylic acid | 93-10-7 | MFCD00006752 | 173.17 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW002011100G
Encompass_Preferred
Quinoline-2-carboxylic acid exhibits antidiabetic activity and can be utilized as a drug intermediate for the synthesis of various active compounds. It reduces the intestinal absorption of carbohydrates by inhibiting α-glucosidase and α-amylase, thereby lowering postprandial blood glucose levels. Additionally, it can serve as a drug intermediate for synthesizing compounds with antiallergic, apoptosis-inducing, or antiproliferative properties.
  • Reduces intestinal absorption of carbohydrates
  • Inhibits α-glucosidase and α-amylase
  • Lowers postprandial blood glucose levels
  • Used for synthesis of antiallergic compounds
  • Used for synthesis of apoptosis-inducing compounds
  • Used for synthesis of antiproliferative compounds
Catalog No. 50-004-47687