Learn More
Medchemexpress LLC Quinoline val-asp difluorophenoxymethylketone | 1135695-98-5 | MFCD08669741 | 99.9% | 513.49 g/mol | C26H25F2N3O6 | 100MG
SDP
Supplier:  Medchemexpress LLC HY12305100MG
Encompass_Preferred
Q-VD-OPh is an irreversible, cell-permeable pan-caspase inhibitor used to block caspase-dependent apoptosis in biochemical and cell-based assays. It inhibits caspase-7 with an IC50 of 48 nM and shows activity against caspases 1, 3, 8, 9, 10, and 12 in the 25-400 nM range. Reported properties include low cytotoxicity and the ability to cross the blood-brain barrier.
  • Potent pan-caspase inhibition with low nanomolar activity.
  • Cell permeable for use in live-cell assays.
  • Low intrinsic cytotoxicity in standard assays.
  • Capable of crossing the blood-brain barrier.
  • High chemical purity suitable for research applications.
  • Supplied as a solid powder for convenient storage and handling.
Catalog No. 50-002-38711