Learn More
Medchemexpress LLC Quinovic acid | 465-74-7 | MFCD01675697 | 486.68 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN15245MG
Encompass_Preferred
Quinovic acid is a triterpene that has demonstrated the ability to mitigate Amyloid-β burden, p53 expression, and cholesterol accumulation. It achieves this by deterring oxidative stress through the upregulation of the Nrf2/HO-1 pathway. Additionally, Quinovic acid can induce apoptosis in cancer cells by upregulating death receptor 5 (DR5). It is used in research concerning cancer, inflammation, metabolic, and neurological diseases, including lung cancer and Alzheimer's disease (AD).
  • Ameliorates Amyloid-β burden, p53 expression, and cholesterol accumulation.
  • Deters oxidative stress by upregulating the Nrf2/HO-1 pathway.
  • Induces apoptosis in cancer cells via upregulation of death receptor 5 (DR5).
  • Applicable for research in various disease areas such as cancer (e.g., lung cancer), inflammation, metabolic, and neurological diseases (e.g., Alzheimer's disease).
Catalog No. 50-004-14865