Learn More
Medchemexpress LLC Quinoxaline, 2,3-di-2-furanyl | 57490-73-0 | MFCD00097796 | 98.3% | 262.26 | C16H10N2O2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY15334910MG
Encompass_Preferred
2,3-Di(furan-2-yl)quinoxaline is a quinoxaline-derived small-molecule fluorescent dye used in laboratory research. It exhibits blue fluorescence, is cell-permeable, and is supplied as a light-yellow solid or as pre-made DMSO solution for convenient use. Manufacturer documentation provides purity, storage, and safety information.
  • Exhibits blue fluorescence suitable for imaging and fluorescence assays.
  • Cell-permeable small molecule for cellular applications.
  • Supplied as a solid and as a 10 mM solution in DMSO for convenience.
  • High purity: 98.3%.
  • Documented storage stability under recommended conditions.
  • Molecular formula C16H10N2O2; molecular weight 262.26.
  • Includes COA and SDS documentation for quality and safety.
Catalog No. 50-002-86171