Learn More
Selleck Chemical LLC Quisinostat (JNJ-26481585) 2HCl 10mM/1mL 875320-31-3
SDP
Supplier:  Selleck Chemical LLC S109610MM/1ML
Encompass_Preferred
Quisinostat (JNJ-26481585) 2HCl is a novel second-generation HDAC inhibitor with highest potency for HDAC1 with IC50 of 0.11 nM in a cell-free assay, modest potent to HDACs 2, 4, 10, and 11; greater than 30-fold selectivity against HDACs 3, 5, 8, and 9 and lowest potency to HDACs 6 and 7. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-6120