Learn More
Medchemexpress LLC Quisinostat dihydrochloride | 875320-31-3 | C21H28Cl2N6O2 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY15433A1MG
Encompass_Preferred
Quisinostat dihydrochloride is an orally active, potent pan-HDAC inhibitor that demonstrates broad-spectrum antitumoral activity and can induce autophagy in neuroblastoma cells.
  • Potent pan-HDAC inhibitor
  • Exhibits broad-spectrum antitumoral activity
  • Induces autophagy in neuroblastoma cells
  • High purity of 99.05%
  • Inhibits HDAC isozymes
  • Displays broad-spectrum antiproliferative activity against solid and hematologic cancer cell lines
  • Induces apoptosis
  • Induces continuous H3 acetylation in tumor tissue in vivo
  • Strongly inhibits the growth of large pre-established HCT116 colon xenografts
Catalog No. 50-003-76851