Learn More
Medchemexpress LLC Quisinostat dihydrochloride | 875320-31-3 | C21H28Cl2N6O2 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY15433A50MG
Encompass_Preferred
Quisinostat dihydrochloride is an orally active, potent pan-HDAC inhibitor with broad-spectrum antitumoral activity. It can induce autophagy in neuroblastoma cells and is intended for research use only.
  • Potent pan-HDAC inhibitor
  • Broad-spectrum antitumoral activity
  • Induces autophagy in neuroblastoma cells
  • Has a purity of 99.05%
  • Appears as a white to yellow solid
Catalog No. 50-003-76190