Learn More
Medchemexpress LLC Quisinostat dihydrochloride (Jnj-26481585 dihydrochloride) | 875320-31-3 | C21H28Cl2N6O2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY15433A100MG
Encompass_Preferred
Quisinostat dihydrochloride is a potent, orally active pan-HDAC (histone deacetylase) inhibitor. It has shown a broad spectrum of antitumoral activity and can induce autophagy in neuroblastoma cells. The product appears as a solid, with a color ranging from white to yellow.
  • Potent pan-HDAC inhibitor
  • Broad spectrum antitumoral activity
  • Induces autophagy
  • High purity (99.05%)
  • Intended for research use only
Catalog No. 50-003-76859