Learn More
Medchemexpress LLC Quizalofop-p-ethyl | 100646-51-3 | 99.1% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB195010MM/1ML
Encompass_Preferred
Quizalofop-p-ethyl is a low-toxicity, highly selective post-emergence herbicide designed for dryland crops. It demonstrates high selectivity between gramineous weeds and dicotyledonous crops, effectively controlling gramineous weeds in broadleaf crop fields.
  • Low-toxicity herbicide.
  • Highly selective post-emergence herbicide for dryland crops.
  • High selectivity between gramineous weeds and dicotyledonous crops.
  • Good control efficacy against gramineous weeds in broadleaf crop fields.
Catalog No. 50-004-12188