Learn More
Electron Microscopy Sciences QuantomiX WetSEM QX -102 Imaging Buffer
SDP
Supplier:  Electron Microscopy Sciences IB64
Encompass_Preferred
Solution optimized for imaging samples in an SEM with QX-102 capsules and is formulated to minimize damage to the samples by the electron beam. Supplied sterile and lyophilized.
Catalog No. 50-313-46