Learn More
Abcam Q x -314 bromide (N-Ethyllidocaine bromide), Na+ channel blocker, 50MG
Supplier:  Abcam AB12011750MG
Encompass_Preferred
MW 343.3 Da, Purity >99%. A membrane impermeable quaternary lidocaine derivative. Blocks voltage-sensitive Na+ conductance when applied intracellularly. Soluble in 1 ml water to give specified mM/ml concentration.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-4685