Learn More
Selleck Chemical LLC QX77
SDP
Supplier:  Selleck Chemical LLC S679725MG
Encompass_Preferred
QX77 is a novel activator of chaperone-mediated autophagy (CMA) QX77 induces the up-regulation of Rab11 expression and up-regulates LAMP2A expression
Catalog No. 50-000-02636