Learn More
Strem, An Ascensus Company CAS# 301847-89-2. 100mg. R-(-)-1,13-Bis(diphenylphosphino)-7,8-dihydro-6H-dibenzo[f,h][1,5]dioxonin, 97% (R)-C3-TUNEPHOS. MFCD06658115
SDP
Supplier:  Strem, An Ascensus Company 150175100MG
Encompass
CAS# 301847-89-2. 100mg. R-(-)-1,13-Bis(diphenylphosphino)-7,8-dihydro-6H-dibenzo[f,h][1,5]dioxonin, 97% (R)-C3-TUNEPHOS. MFCD06658115. Molecular Weight: 594.62. Molecular Formula: C39H32O2P2. Color/form: white pwdr. Strem# 15-0175. http://www.strem.com/catalog/v/15-0175/
Catalog No. 50-901-09822